1-Brom-2-fluor-4-(trifluormethoxy)benzol

Lieferant: COMBI-BLOCKS

OS-7663-5G OS-7663-25G OS-7663-1G
COBBOS-7663-5GEA 48.4 EUR
COBBOS-7663-5G COBBOS-7663-25G COBBOS-7663-1G
1-Brom-2-fluor-4-(trifluormethoxy)benzol
1-Bromo-2-fluoro-4-(trifluoromethoxy)benzene
Formel: C₇H₃BrF₄O
Molecular Weight: 259.00 g/mol
MDL: MFCD06657548
CAS-Nummer: 168971-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR