1-Brom-2-fluor-5-(trifluormethoxy)benzol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Bromo-1-fluoro-4-(trifluoromethoxy)benzene

H32650.03 H32650.06 H32650.14
H32650.03EA 20.2 EUR
H32650.03 H32650.06 H32650.14
1-Brom-2-fluor-5-(trifluormethoxy)benzol ≥98%
1-Bromo-2-fluoro-5-(trifluoromethoxy)benzene

Purity: 98% CAS No.: 286932-57-8 Molecular Formula: C7H3BrF4O Formula Weight: 259.00 Physical Form: Liquid Appearance: Colourless to yellow MDL No.: MFCD02181186

Formel: C₇H₃BrF₄O
Molecular Weight: 259,00 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02181186
CAS-Nummer: 286932-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR