1-Brom-3-(2,2,2-trifluorethoxy)benzol

Lieferant: COMBI-BLOCKS

OT-0432-1G OT-0432-5G
COBBOT-0432-1GEA 204 EUR
COBBOT-0432-1G COBBOT-0432-5G
1-Brom-3-(2,2,2-trifluorethoxy)benzol
1-Bromo-3-(2,2,2-trifluoroethoxy)benzene
Formel: C₈H₆BrF₃O
MDL: MFCD09027280
CAS-Nummer: 888327-41-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR