4-Brom-2-fluor-1-(2-methoxyethoxy)benzol

Lieferant: COMBI-BLOCKS

OT-1161-5G OT-1161-1G
COBBOT-1161-5GEA 1180 EUR
COBBOT-1161-5G COBBOT-1161-1G
4-Brom-2-fluor-1-(2-methoxyethoxy)benzol
4-Brom-2-fluor-1-(2-methoxyethoxy)benzol
Formel: C₉H₁₀BrFO₂
MDL: MFCD15143588
CAS-Nummer: 944279-01-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR