1-Brom-4-fluor-2-isopropoxybenzol

Lieferant: COMBI-BLOCKS

OT-0684-5G OT-0684-1G
COBBOT-0684-5GEA 434 EUR
COBBOT-0684-5G COBBOT-0684-1G
1-Brom-4-fluor-2-isopropoxybenzol
1-Brom-4-fluor-2-isopropoxybenzol
Formel: C₉H₁₀BrFO
MDL: MFCD10699697
CAS-Nummer: 610797-49-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR