1-Brom-2-isopropoxybenzol ≥97%
Lieferant: Thermo Scientific
Synonyme:
2-(2'-Bromophenoxy)propane
B25688.06
B25688.14
B25688.06EA
42.5
EUR
B25688.06
B25688.14
1-Brom-2-isopropoxybenzol ≥97%
1-Brom-2-isopropoxybenzol
Purity: 97%
CAS No.: 701-07-5
Molecular Formula: C9H11BrO
Formula Weight: 215.09
Odour: Aromatic
Boiling Point: 73°/2mm
MDL No.: MFCD00070759
Formel:
C₉H₁₁BrO Molecular Weight: 215,09 g/mol Siedepunkt: 73 °C (2 mmHg) Dichte: 1,34 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00070759 CAS-Nummer: 701-07-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...