4-Chlor-2-ethoxy-1-nitrobenzol

Lieferant: COMBI-BLOCKS

AN-1057-5G AN-1057-1G
COBBAN-1057-5GEA 0 EUR
COBBAN-1057-5G COBBAN-1057-1G
4-Chlor-2-ethoxy-1-nitrobenzol
4-Chlor-2-ethoxy-1-nitrobenzol
Formel: C₈H₈ClNO₃
MDL: MFCD02625956
CAS-Nummer: 29604-25-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR