3-(Methylamino)benzotrifluorid

Lieferant: COMBI-BLOCKS

Synonyme: N-Methyl-3-(trifluoromethyl)aniline

HC-2918-5G HC-2918-1G
COBBHC-2918-5GEA 324 EUR
COBBHC-2918-5G COBBHC-2918-1G
3-(Methylamino)benzotrifluorid
3-(Methylamino)benzotrifluorid
Formel: C₈H₈F₃N
Molecular Weight: 175,15 g/mol
MDL: MFCD07368696
CAS-Nummer: 2026-70-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR