4-(Triethylsilyl)-3-butin-1-ol

Lieferant: CHEMPUR SILANE

Synonyme: 4-(Triethylsilyl)but-3-yn-1-ol

SILS17618.5 SILS17618.25
SILNSILS17618.5EA 0 EUR
SILNSILS17618.5 SILNSILS17618.25
4-(Triethylsilyl)-3-butin-1-ol
4-(Triethylsilyl)-3-butin-1-ol
Formel: C₁₀H₂₀OSi
Molecular Weight: 184,35 g/mol
MDL: MFCD00190219
CAS-Nummer: 160194-29-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR