2-Chlor-5-propoxyphenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (2-Chloro-5-propoxyphenyl)boronic acid

BB-6389-5G BB-6389-1G
COBBBB-6389-5GEA 911 EUR
COBBBB-6389-5G COBBBB-6389-1G
2-Chlor-5-propoxyphenylboronsäure
2-Chlor-5-propoxyphenylboronsäure
Formel: C₉H₁₂BClO₃
Molecular Weight: 214.4538 g/mol
MDL: MFCD16295074
CAS-Nummer: 1256346-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR