3-Chlor-4-propoxyphenylboronsäure
Lieferant: COMBI-BLOCKS
BB-8135-1G
BB-8135-250M
COBBBB-8135-1GEA
75.9
EUR
COBBBB-8135-1G
COBBBB-8135-250M
3-Chlor-4-propoxyphenylboronsäure
3-Chlor-4-propoxyphenylboronsäure
Formel:
C₉H₁₂BClO₃ |
MDL:
MFCD03701555 CAS-Nummer: 480438-57-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...