5-Brom-2-fluor-4-methylanilin

Lieferant: COMBI-BLOCKS

HC-2065-5G HC-2065-1G
COBBHC-2065-5GEA 449 EUR
COBBHC-2065-5G COBBHC-2065-1G
5-Brom-2-fluor-4-methylanilin
5-Brom-2-fluor-4-methylanilin
Formel: C₇H₇BrFN
Molecular Weight: 204,0400 g/mol
MDL: MFCD11504886
CAS-Nummer: 945244-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR