4-Brom-2-fluor-5-methylanilin

Lieferant: CHEMPUR

FL13744.POR 013744-25G 013744-5G
CHMPFL13744.POREA 0 EUR
CHMPFL13744.POR FLUO013744-25G FLUO013744-5G
4-Brom-2-fluor-5-methylanilin
4-Brom-2-fluor-5-methylanilin
Formel: C₇H₇BrFN
Molecular Weight: 204,0400 g/mol
MDL: MFCD04973744
CAS-Nummer: 418762-26-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR