4-Brom-5-fluor-2-methylanilin

Lieferant: CHEMPUR

FL13781.POR 013781-25G 013781-5G
CHMPFL13781.POREA 0 EUR
CHMPFL13781.POR FLUO013781-25G FLUO013781-5G
4-Brom-5-fluor-2-methylanilin
4-Brom-5-fluor-2-methylanilin
Formel: C₇H₇BrFN
Molecular Weight: 204,0400 g/mol
MDL: MFCD04973753
CAS-Nummer: 52723-82-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR