1-Brom-2-(methoxymethyl)benzol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromobenzyl methyl ether

OT-0534-1G OT-0534-5G
COBBOT-0534-1GEA 56.9 EUR
COBBOT-0534-1G COBBOT-0534-5G
1-Brom-2-(methoxymethyl)benzol
1-Bromo-2-(methoxymethyl)benzene
Formel: C₈H₉BrO
Molecular Weight: 201,0605 g/mol
MDL: MFCD03790887
CAS-Nummer: 52711-30-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR