1-Brom-3-(methoxymethyl)benzol
Lieferant: COMBI-BLOCKS
OT-0766-5G
OT-0766-25G
OT-0766-1G
COBBOT-0766-5GEA
423
EUR
COBBOT-0766-5G
COBBOT-0766-25G
COBBOT-0766-1G
1-Brom-3-(methoxymethyl)benzol
1-Bromo-3-(methoxymethyl)benzene
Formel:
C₈H₉BrO Molecular Weight: 201.0600 g/mol |
MDL:
MFCD11504955 CAS-Nummer: 1515-89-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...