3-Ethylbenzonitril

Lieferant: CHEMPUR

TL00572.1 TL00572.10
CHMPTL00572.1EA 0 EUR
CHMPTL00572.1 CHMPTL00572.10
3-Ethylbenzonitril
3-Ethylbenzonitril
Formel: C₉H₉N
Molecular Weight: 131.1700 g/mol
MDL: MFCD00045601
CAS-Nummer: 34136-57-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR