4-Butylbiphenyl

Lieferant: Thermo Scientific

Synonyme: 4-Butyl-1,1'-Biphenyl

BTB10237.1 BTB10237.10 BTB10237.50
MAYBBTB10237.1EA 0 EUR
MAYBBTB10237.1 MAYBBTB10237.10 MAYBBTB10237.50
4-Butylbiphenyl
4-Butylbiphenyl
Formel: C₁₆H₁₈
Molecular Weight: 210,32 g/mol
MDL: MFCD00100665
CAS-Nummer: 37909-95-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR