5-tert-Butyl-2-iod-1,3-dimethylbenzol ≥97%
Lieferant: Thermo Scientific
B25195.03 B25195.06
B25195.03EA 68.5 EUR
B25195.03 B25195.06
5-tert-Butyl-2-iod-1,3-dimethylbenzol ≥97%
5-tert-Butyl-2-iod-1,3-dimethylbenzol
Purity: 97% CAS No.: 5122-20-3 Molecular Formula: C12H17I Formula Weight: 288.17 Odour: Odourless Melting Point: 54-57° MDL No.: MFCD00052173
Formel: C₁₂H₁₇I
Molecular Weight: 288,17 g/mol
Schmelzpunkt: 54…57 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052173
CAS-Nummer: 5122-20-3