5,7-Dichlorchinolin

Lieferant: COMBI-BLOCKS

HC-6979-1G HC-6979-5G
COBBHC-6979-1GEA 149 EUR
COBBHC-6979-1G COBBHC-6979-5G
5,7-Dichlorchinolin
5,7-Dichloroquinoline
Formel: C₉H₅Cl₂N
Molecular Weight: 198,0500 g/mol
MDL: MFCD11504933
CAS-Nummer: 4964-77-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR