8-(Trifluormethyl)-4-chinolinol

Lieferant: COMBI-BLOCKS

HC-6483-5G HC-6483-1G
COBBHC-6483-5GEA 0 EUR
COBBHC-6483-5G COBBHC-6483-1G
8-(Trifluormethyl)-4-chinolinol
8-(Trifluormethyl)-4-chinolinol
Formel: C₁₀H₆F₃NO
MDL: MFCD00134577
CAS-Nummer: 23779-96-6
EINECS: 245-879-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR