6-Amino-2-methylchinolin 97%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
2-methylquinolin-6-amine
A13967.03
A13967.06
A13967.03EA
84.9
EUR
A13967.03
A13967.06
6-Amino-2-methylchinolin 97%
6-Amino-2-methylquinoline
Purity: 97%
CAS No.: 65079-19-8
Molecular Formula: C10H10N2
Formula Weight: 158.20
Melting Point: 189-191°
Beilstein Reference: 114795
Formel:
C₁₀H₁₀N₂ Molecular Weight: 158,20 g/mol Schmelzpunkt: 189…191 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052600 CAS-Nummer: 65079-19-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...