6-Acetamido-3-chlor-2-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Acetamido-3-chloro-2-picoline

PY-8024-1G PY-8024-5G
COBBPY-8024-1GEA 154 EUR
COBBPY-8024-1G COBBPY-8024-5G
6-Acetamido-3-chlor-2-methylpyridin
6-Acetamido-3-chlor-2-methylpyridin
Formel: C₈H₉ClN₂O
MDL: MFCD08713015
CAS-Nummer: 160115-16-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR