2-Acetylamino-4-methylpyridin
Lieferant: COMBI-BLOCKS
Synonyme:
N-(4-Methyl-2-pyridyl)acetamide, N-(4-Methylpyridin-2-yl)acetamide, 2-Acetamido-4-picoline, 2-Acetamido-4-methylpyridine
CA-4089-5G
CA-4089-25G
CA-4089-1G
COBBCA-4089-5GEA
102
EUR
COBBCA-4089-5G
COBBCA-4089-25G
COBBCA-4089-1G
2-Acetylamino-4-methylpyridin
2-Acetylamino-4-methylpyridin
Formel:
C₈H₁₀N₂O Molecular Weight: 150.1778 g/mol |
MDL:
MFCD00234432 CAS-Nummer: 5327-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...