tert-Butyl-5-brompicolinat
Lieferant: COMBI-BLOCKS
Synonyme:
tert-Butyl 5-bromopicolinate, 5-Bromo-pyridine-2-carboxylic acid t-butyl ester, tert-Butyl 5-bromopyridine-2-carboxylate
CA-4227-5G
CA-4227-1G
COBBCA-4227-5GEA
711
EUR
COBBCA-4227-5G
COBBCA-4227-1G
tert-Butyl-5-brompicolinat
tert-Butyl-5-brompicolinat
Formel:
C₁₀H₁₂BrNO₂ Molecular Weight: 258.1100 g/mol |
MDL:
MFCD08436052 CAS-Nummer: 845306-08-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...