Diethoxymethylsilan

Lieferant: CHEMPUR SILANE

Synonyme: Diethoxy(methyl)silane

SILS11225.10 SILS11225.50 SILS11225.100
SILNSILS11225.10EA 0 EUR
SILNSILS11225.10 SILNSILS11225.50 SILNSILS11225.100
Diethoxymethylsilan
Diethoxymethylsilan
Formel: C₅H₁₄O₂Si
Molecular Weight: 134,25 g/mol
MDL: MFCD00026749
CAS-Nummer: 2031-62-1
EINECS: 217-982-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR