1-Aminopiperidin 97%

Lieferant: Thermo Fisher Scientific
Gefahr

104410100 104410250
ACRO104410100EA 90 EUR
ACRO104410100 ACRO104410250
1-Aminopiperidin 97%
1-Aminopiperidin
Formel: C₅H₁₂N₂
Molecular Weight: 100.16 g/mol
Dichte: 0.928 g/cm³
Flammpunkt: 36 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006489
CAS-Nummer: 2213-43-6
EINECS: 218-666-8
UN: 2920
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4735 to 1.4775 (20°C, 589 nm)
GC ≥96.0 %
Calcium (Ca) ≤200 ppm
Copper (Cu) ≤10 ppm
Iron (Fe) ≤200 ppm
Insoluble matter ≤1 % (in acid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR