2-(2-Bromethyl)-1,3-dioxan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

269131-10G 269131-50G
SIAL269131-10GEA 55.8 EUR
SIAL269131-10G SIAL269131-50G
2-(2-Bromethyl)-1,3-dioxan, Sigma-Aldrich®
2-(2-Bromethyl)-1,3-dioxan
Formel: C₆H₁₁BrO₂
Molecular Weight: 195,06 g/mol
MDL: MFCD00006567
CAS-Nummer: 33884-43-4
EINECS: 251-716-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR