2-(2,4-Dichlorphenoxy)benzyl alcohol

Lieferant: BIONET RESEARCH

2P-077.500 2P-077.1
BIOT2P-077.500EA 0 EUR
BIOT2P-077.500 BIOT2P-077.1
2-(2,4-Dichlorphenoxy)benzyl alcohol
2-(2,4-Dichlorphenoxy)benzyl alcohol
Formel: C₁₃H₁₀Cl₂O₂
MDL: MFCD01814784
CAS-Nummer: 361212-70-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR