1-(4-Bromphenyl)piperidin

Lieferant: COMBI-BLOCKS

HC-2492-1G HC-2492-5G
COBBHC-2492-1GEA 83.6 EUR
COBBHC-2492-1G COBBHC-2492-5G
1-(4-Bromphenyl)piperidin
1-(4-Bromphenyl)piperidin
Formel: C₁₁H₁₄BrN
Molecular Weight: 240,14 g/mol
MDL: MFCD04112533
CAS-Nummer: 22148-20-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR