2-Brom-4'-(1-piperidinyl)acetophenon

Lieferant: COMBI-BLOCKS

Synonyme: 4-(Piperidin-1-yl)phenacyl bromide

OT-0493-5G OT-0493-1G
COBBOT-0493-5GEA 0 EUR
COBBOT-0493-5G COBBOT-0493-1G
2-Brom-4'-(1-piperidinyl)acetophenon
2-Brom-4'-(1-piperidinyl)acetophenon
Formel: C₁₃H₁₆BrNO
MDL: MFCD06808683
CAS-Nummer: 210832-84-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR