1-(3,4-Dichlorphenyl)pyrrolidin

Lieferant: COMBI-BLOCKS

HC-6388-5G HC-6388-1G
COBBHC-6388-5GEA 303 EUR
COBBHC-6388-5G COBBHC-6388-1G
1-(3,4-Dichlorphenyl)pyrrolidin
1-(3,4-Dichlorphenyl)pyrrolidin
Formel: C₁₀H₁₁Cl₂N
Molecular Weight: 216.1 g/mol
MDL: MFCD09878388
CAS-Nummer: 957065-92-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR