5-(Trifluormethyl)picolinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 5-(Trifluoromethyl)pyridine-2-carboxylic acid

PY-1447-1G PY-1447-25G PY-1447-5G
COBBPY-1447-1GEA 26.2 EUR
COBBPY-1447-1G COBBPY-1447-25G COBBPY-1447-5G
5-(Trifluormethyl)picolinsäure
5-(Trifluormethyl)picolinsäure
Formel: C₇H₄F₃NO₂
Molecular Weight: 191,11 g/mol
MDL: MFCD04113632
CAS-Nummer: 80194-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR