6-(Trifluormethyl)picolinsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-(Trifluoromethyl)-2-pyridinecarboxylic acid, 6-(Trifluoromethyl)picolinic acid, 2-(Trifluoromethyl)-6-pyridinecarboxylic acid, 6-(Trifluoromethyl)pyridine-2-carboxylic acid
H63138.MD
H63138.03
H63138.MDEA
41.7
EUR
H63138.MD
H63138.03
6-(Trifluormethyl)picolinsäure ≥98%
6-(Trifluormethyl)picolinsäure
Purity: 98%
CAS No.: 131747-42-7
Molecular Formula: C7H4F3NO2
Formula Weight: 191.11
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 154-158°
UN No.: 2811
MDL No.: MFCD02180819
Formel:
C₇H₄F₃NO₂ Molecular Weight: 191,11 g/mol Schmelzpunkt: 154…158 °C |
MDL:
MFCD02180819 CAS-Nummer: 131747-42-7 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...