1-(Heptafluorbutyryl)imidazol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
1-(Perfluorobutanoyl)-1H-imidazole, N-Heptafluorobutyrylimidazole
M-Clarity™ Qualitätsstufe = MQ100
556645-1G
556645-5G
SIAL556645-1GEA
45.2
EUR
SIAL556645-1G
SIAL556645-5G
1-(Heptafluorbutyryl)imidazol, Sigma-Aldrich®
1-(Heptafluorbutyryl)imidazol
Formel:
C₇H₃F₇N₂O Molecular Weight: 264.10 g/mol |
MDL:
MFCD00014503 CAS-Nummer: 32477-35-3 EINECS: 251-063-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...