2,4-Dibrom-1-tritylimidazol

Lieferant: COMBI-BLOCKS

HI-1151-5G HI-1151-1G
COBBHI-1151-5GEA 1460 EUR
COBBHI-1151-5G COBBHI-1151-1G
2,4-Dibrom-1-tritylimidazol
2,4-Dibrom-1-tritylimidazol
Formel: C₂₂H₁₆Br₂N₂
MDL: MFCD07363849
CAS-Nummer: 871269-06-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR