4,5-Dibrom-1-tritylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 4,5-Dibromo-1-triphenylmethyl-1H-imidazole

HI-1153-1G HI-1153-5G
COBBHI-1153-1GEA 315 EUR
COBBHI-1153-1G COBBHI-1153-5G
4,5-Dibrom-1-tritylimidazol
4,5-Dibrom-1-tritylimidazol
Formel: C₂₂H₁₆Br₂N₂
MDL: MFCD08056373
CAS-Nummer: 112517-23-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR