4-Brom-1-tritylimidazol

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-1-trityl-1H-imidazole

HI-1084-5G HI-1084-1G
COBBHI-1084-5GEA 591 EUR
COBBHI-1084-5G COBBHI-1084-1G
4-Brom-1-tritylimidazol
4-Brom-1-tritylimidazol
Formel: C₂₂H₁₇BrN₂
Molecular Weight: 389,2878 g/mol
MDL: MFCD00185657
CAS-Nummer: 87941-55-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR