4-Iod-1-tritylimidazol
Lieferant: COMBI-BLOCKS
Synonyme:
4-Iodo-1-trityl-1H-imidazole
HI-1064-5G
HI-1064-1G
COBBHI-1064-5GEA
122
EUR
COBBHI-1064-5G
COBBHI-1064-1G
4-Iod-1-tritylimidazol
4-Iod-1-tritylimidazol
Formel:
C₂₂H₁₇IN₂ Molecular Weight: 436.2900 g/mol |
MDL:
MFCD02179542 CAS-Nummer: 96797-15-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...