2-Benzyloxy-1-chlor-4-methylbenzol

Lieferant: COMBI-BLOCKS

OT-0709-1G OT-0709-5G
COBBOT-0709-1GEA 149 EUR
COBBOT-0709-1G COBBOT-0709-5G
2-Benzyloxy-1-chlor-4-methylbenzol
2-Benzyloxy-1-chlor-4-methylbenzol
Formel: C₁₄H₁₃ClO
MDL: MFCD11053830
CAS-Nummer: 1065074-77-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR