Kalium(4-methyl-1-naphthyl)trifluoroborat

Lieferant: COMBI-BLOCKS

TB-3218-1G TB-3218-5G TB-3218-25G
COBBTB-3218-1GEA 50.9 EUR
COBBTB-3218-1G COBBTB-3218-5G COBBTB-3218-25G
Kalium(4-methyl-1-naphthyl)trifluoroborat
Potassium(4-methyl-1-naphthalene)trifluoroborate
Formel: C₁₁H₉BF₃K
Molecular Weight: 248,09 g/mol
MDL: MFCD04115748
CAS-Nummer: 850623-55-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR