Dikalium-1,4-phenylenbis(trifluoroborat)

Lieferant: COMBI-BLOCKS

TB-2630-1G TB-2630-5G
COBBTB-2630-1GEA 215 EUR
COBBTB-2630-1G COBBTB-2630-5G
Dikalium-1,4-phenylenbis(trifluoroborat)
Dipotassium phenylene-1,4-bistrifluoroborate
Formel: C₆H₄B₂F₆K₂
MDL: MFCD12026116
CAS-Nummer: 1150655-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR