4-(4-Bromphenoxy)tetrahydropyran

Lieferant: COMBI-BLOCKS

Synonyme: 4-(4-Bromophenoxy)tetrahydro-2H-pyran

HC-2263-5G HC-2263-1G HC-2263-250M
COBBHC-2263-5GEA 0 EUR
COBBHC-2263-5G COBBHC-2263-1G COBBHC-2263-250M
4-(4-Bromphenoxy)tetrahydropyran
4-(4-Bromphenoxy)tetrahydropyran
Formel: C₁₁H₁₃BrO₂
MDL: MFCD11855868
CAS-Nummer: 215453-84-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR