2-Acetyl-3-thiophenboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
2-Acetyl-3-thiopheneboronic acid, 2-Acetyl-3-thienylboronic acid, (2-Acetylthiophen-3-yl)boronic acid
BB-9084-5G
BB-9084-1G
COBBBB-9084-5GEA
474
EUR
COBBBB-9084-5G
COBBBB-9084-1G
2-Acetyl-3-thiophenboronsäure
2-Acetyl-3-thiophenboronsäure
Formel:
C₆H₇BO₃S Molecular Weight: 169,9940 g/mol |
MDL:
MFCD01318932 CAS-Nummer: 36155-74-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...