4-Amino-3,5-diphenyl-1,2,4-triazol

Lieferant: BIONET RESEARCH

Synonyme: Diphenyl-4H-1,2,4-triazol-4-amine, 3,5-Diphenyl-4H-1,2,4-triazol-4-amine

8P-933.500 8P-933.1 8P-933.5
BIOT8P-933.500EA 0 EUR
BIOT8P-933.500 BIOT8P-933.1 BIOT8P-933.5
4-Amino-3,5-diphenyl-1,2,4-triazol
4-Amino-3,5-diphenyl-1,2,4-triazole
Formel: C₁₄H₁₂N₄
Molecular Weight: 236,2700 g/mol
MDL: MFCD00450576
CAS-Nummer: 3049-45-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR