Ethyl-2-piperidincarboxylat 98+%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Ethyl 2-Piperidinecarboxylate, Ethyl pipecolinate

189580050 189580250
ACRO189580050EA 45.6 EUR
ACRO189580050 ACRO189580250
Ethyl-2-piperidincarboxylat 98+%
Ethyl-2-piperidincarboxylat
Formel: C₈H₁₅NO₂
Molecular Weight: 157.21 g/mol
Dichte: 1.006 g/cm³
Flammpunkt: 46 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005980
CAS-Nummer: 15862-72-3
EINECS: 239-990-6
UN: 1993
ADR: 3 III

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4545 to 1.4575 (20°C, 589 nm)
GC ≥98.0 %

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR