Ethyl-3-piperidincarboxylat 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ethyl nipecotate, Ethyl 3-Piperidinecarboxylate, Ethyl piperidine-3-carboxylate
186620250
186621000
186625000
ACRO186620250EA
36.9
EUR
ACRO186620250
ACRO186621000
ACRO186625000
Ethyl-3-piperidincarboxylat 97%
Ethyl-3-piperidincarboxylat
Formel:
C₈H₁₅NO₂ Molecular Weight: 157.21 g/mol Siedepunkt: 102…104 °C (9hPa) Dichte: 1.012 g/cm³ Flammpunkt: 90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005991 CAS-Nummer: 5006-62-2 EINECS: 225-681-3 ADR: 8 III |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow-brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4580 to 1.4610 (20°C, 589 nm) |
GC | ≥96.0 % |
HPLC | ≥98.5 % |
Residue after ignition | ≤0.1 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...