Ethyl-3-piperidincarboxylat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl nipecotate, Ethyl 3-Piperidinecarboxylate, Ethyl piperidine-3-carboxylate
009838-5G
009838-25G
009838-100G
FLUO009838-5GEA
0
EUR
FLUO009838-5G
FLUO009838-25G
FLUO009838-100G
Ethyl-3-piperidincarboxylat
Ethyl-3-piperidincarboxylat
Formel:
C₈H₁₅NO₂ Molecular Weight: 157.21 g/mol |
MDL:
MFCD00005991 CAS-Nummer: 5006-62-2 EINECS: 225-681-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...