3-[(Trifluormethyl)thio]anilin 96%

Lieferant: Thermo Scientific
Achtung

B23734.03 B23734.06
B23734.03EA 68.9 EUR
B23734.03 B23734.06
3-[(Trifluormethyl)thio]anilin 96%
3-[(Trifluormethyl)thio]anilin

Purity: 96% CAS No.: 369-68-6 Molecular Formula: C7H6F3NS Formula Weight: 193.19 Flash Point: 103°(218°F) Refractive Index: 1.5220 Odour: Unpleasant Boiling Point: 221-224° UN No.: 2810 MDL No.: MFCD00190130

Formel: C₇H₆F₃NS
Molecular Weight: 193,19 g/mol
Siedepunkt: 221…224 °C
Dichte: 1,348
Flammpunkt: 103 °C (218 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00190130
CAS-Nummer: 369-68-6
EINECS: 206-722-4
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR