3-Cyclopenten-1-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
cyclopent-3-ene-1-carboxylic acid, Cyclopent-3-enecarboxylic acid
011660-5G
011660-25G
FLUO011660-5GEA
36.9
EUR
FLUO011660-5G
FLUO011660-25G
3-Cyclopenten-1-carbonsäure
3-Cyclopenten-1-carbonsäure
Formel:
C₆H₈O₂ Molecular Weight: 112.13 g/mol |
MDL:
MFCD00101764 CAS-Nummer: 7686-77-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...